Anti-Growth Hormone Mouse Monoclonal Antibody [clone: GH-2]

Proveedor: Abcam
ABCAAB9822-100EA 799 EUR
ABCAAB9822-100
Anti-Growth Hormone Mouse Monoclonal Antibody [clone: GH-2]
Anticuerpos
Type: Primary
Antigen: Growth Hormone
Clonality: Monoclonal
Clone: GH-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR