Anti-GDNF Rabbit Polyclonal Antibody

ABCAAB9789-100EA 611 EUR
ABCAAB9789-100
Anti-GDNF Rabbit Polyclonal Antibody
Type: Primary
Antigen: GDNF (glial cell derived neurotrophic factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR