Anti-GRO beta Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9777-100EA 799 EUR
ABCAAB9777-100
Anti-GRO beta Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: GRO beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR