Anti-IL1A Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9724-100EA 756 EUR
ABCAAB9724-100
Anti-IL1A Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: IL1A (interleukin 1, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR