Anti-AMPD1 Goat Polyclonal Antibody

ABCAAB9699-100EA 588 EUR
ABCAAB9699-100
Anti-AMPD1 Goat Polyclonal Antibody
Type: Primary
Antigen: AMPD1 (adenosine monophosphate deaminase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR