Anti-CSF3 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9691-100EA 560 EUR
ABCAAB9691-100
Anti-CSF3 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CSF3 (colony stimulating factor 3 (granulocyte))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR