Anti-XCL1 Rabbit Polyclonal Antibody

ABCAAB9686-100EA 754 EUR
ABCAAB9686-100
Anti-XCL1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: XCL1 (chemokine (C motif) ligand 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR