Anti-Interferon gamma Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9657-100EA 820 EUR
ABCAAB9657-100
Anti-Interferon gamma Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Interferon gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR