Anti-Retinoic Acid Receptor alpha Mouse Monoclonal Antibody [clone: 763]

ABCAAB9639-50EA 0 EUR
ABCAAB9639-50
Anti-Retinoic Acid Receptor alpha Mouse Monoclonal Antibody [clone: 763]
Type: Primary
Antigen: Retinoic Acid Receptor alpha
Clonality: monoclonal
Clone: 763
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR