Anti-IL2 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9618-100EA 550 EUR
ABCAAB9618-100
Anti-IL2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: IL2 (interleukin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR