Anti-TGF alpha Mouse Monoclonal Antibody [clone: P/T1]

Proveedor: Abcam
ABCAAB9578-100EA 0 EUR
ABCAAB9578-100
Anti-TGF alpha Mouse Monoclonal Antibody [clone: P/T1]
Anticuerpos
Type: Primary
Antigen: TGF alpha
Clonality: monoclonal
Clone: P/T1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR