Anti-CD74 Mouse Monoclonal Antibody [clone: LN2]

Proveedor: Abcam
ABCAAB9514-500EA 585 EUR
ABCAAB9514-500
Anti-CD74 Mouse Monoclonal Antibody [clone: LN2]
Anticuerpos
Type: Primary
Antigen: CD74 (CD74 molecule, major histocompatibility complex, class II invariant chain)
Clonality: monoclonal
Clone: LN2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR