Anti-GAPDH Mouse Monoclonal Antibody [clone: mAbcam 9484]

ABCAAB9484-200EA 681 EUR
ABCAAB9484-200
Anti-GAPDH Mouse Monoclonal Antibody [clone: mAbcam 9484]
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: monoclonal
Clone: mAbcam 9484
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR