Anti-GAPDH Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9483-100EA 789 EUR
ABCAAB9483-100
Anti-GAPDH Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR