Anti-MS4A1 Mouse Monoclonal Antibody [clone: L26]

Proveedor: Abcam
ABCAAB9475-250EA 0 EUR
ABCAAB9475-250
Anti-MS4A1 Mouse Monoclonal Antibody [clone: L26]
Anticuerpos
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone: L26
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR