Anti-AR Mouse Monoclonal Antibody [clone: AR 441]

Proveedor: Abcam
ABCAAB9474-250EA 894 EUR
ABCAAB9474-250
Anti-AR Mouse Monoclonal Antibody [clone: AR 441]
Anticuerpos
Type: Primary
Antigen: AR (androgen receptor)
Clonality: monoclonal
Clone: AR 441
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR