Anti-L-Aspartate Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9439-500EA 793 EUR
ABCAAB9439-500
Anti-L-Aspartate Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: L-Aspartate
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR