Anti-SERPINA1 Mouse Monoclonal Antibody [clone: G11]

Proveedor: Abcam
ABCAAB9400-100EA 854 EUR
ABCAAB9400-100
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: G11]
Anticuerpos
Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: monoclonal
Clone: G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR