Anti-SERPINA1 Mouse Monoclonal Antibody [clone: B9]

Proveedor: Abcam
ABCAAB9399-100EA 999 EUR
ABCAAB9399-100
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: B9]
Anticuerpos
Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: monoclonal
Clone: B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR