Anti-GAPDH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Abcam
ABCAAB9385-100EA 530 EUR
ABCAAB9385-100
Anti-GAPDH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now