Anti-Cytokeratin 10 + 13 Mouse Monoclonal Antibody [clone: DE-K13]

Proveedor: Abcam
ABCAAB9383-100EA 813 EUR
ABCAAB9383-100
Anti-Cytokeratin 10 + 13 Mouse Monoclonal Antibody [clone: DE-K13]
Anticuerpos
Type: Primary
Antigen: Cytokeratin 10 + 13
Clonality: monoclonal
Clone: DE-K13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR