Anti-SERPINA1 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9373-500EA 815 EUR
ABCAAB9373-500
Anti-SERPINA1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR