Anti-S6K Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Abcam
ABCAAB9367-100EA 787 EUR
ABCAAB9367-100
Anti-S6K Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Primary
Antigen: S6K
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR