Anti-S6K Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9366-100EA 0 EUR
ABCAAB9366-100
Anti-S6K Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: S6K
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR