Anti-Macrophage Inflammatory Protein 3 alpha Mouse Monoclonal Antibody

ABCAAB9349-500EA 611 EUR
ABCAAB9349-500
Anti-Macrophage Inflammatory Protein 3 alpha Mouse Monoclonal Antibody
Type: Primary
Antigen: Macrophage Inflammatory Protein 3 alpha
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR