Anti-TNF Mouse Monoclonal Antibody

Proveedor: Abcam
ABCAAB9348-500EA 838 EUR
ABCAAB9348-500
Anti-TNF Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR