Anti-Phosphothreonine Rabbit Polyclonal Antibody (Biotin)
Proveedor: Abcam
ABCAAB9340-100EA 838 EUR
ABCAAB9340-100
Anti-Phosphothreonine Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: Phosphothreonine
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now