Anti-Phosphoserine Rabbit Polyclonal Antibody (Biotin)

Proveedor: Abcam
ABCAAB9335-50EA 848 EUR
ABCAAB9335-50
Anti-Phosphoserine Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: Phosphoserine
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR