Anti-KRT1 Mouse Monoclonal Antibody [clone: AE1]
Proveedor: Abcam
ABCAAB9286-125EA 881 EUR
ABCAAB9286-125
Anti-KRT1 Mouse Monoclonal Antibody [clone: AE1]
Anticuerpos
Type: Primary
Antigen: KRT1 (keratin 1)
Clonality: monoclonal
Clone: AE1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now