Anti-NCAM1 Mouse Monoclonal Antibody [clone: 123C3]

Proveedor: Abcam
ABCAAB9272-100EA 828 EUR
ABCAAB9272-100
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 123C3]
Anticuerpos
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone: 123C3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR