Anti-KIF23 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9259-50EA 550 EUR
ABCAAB9259-50
Anti-KIF23 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: KIF23 (kinesin family member 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR