Anti-RFX5 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9255-50EA 849 EUR
ABCAAB9255-50
Anti-RFX5 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: RFX5 (regulatory factor X, 5 (influences HLA class II expression))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR