Anti-M13 Bacteriophage Coat Protein g8p Mouse Monoclonal Antibody [clone: RL-ph2]

Proveedor: Abcam
ABCAAB9225-100EA 814 EUR
ABCAAB9225-100
Anti-M13 Bacteriophage Coat Protein g8p Mouse Monoclonal Antibody [clone: RL-ph2]
Anticuerpos
Type: Primary
Antigen: M13 Bacteriophage Coat Protein g8p
Clonality: monoclonal
Clone: RL-ph2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR