Anti-M13 Bacteriophage Coat Protein g8p Mouse Monoclonal Antibody [clone: RL-ph1]

ABCAAB9223-100EA 651 EUR
ABCAAB9223-100
Anti-M13 Bacteriophage Coat Protein g8p Mouse Monoclonal Antibody [clone: RL-ph1]
Type: Primary
Antigen: M13 Bacteriophage Coat Protein g8p
Clonality: monoclonal
Clone: RL-ph1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR