Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Horse Polyclonal Antibody

Proveedor: Abcam
ABCAAB9193-500EA 0 EUR
ABCAAB9193-500
Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Horse Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Hepatitis B Virus Surface Antigen (Ad/Ay)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Horse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR