Anti-IgM Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9175-1EA 340 EUR
ABCAAB9175-1
Anti-IgM Rabbit Polyclonal Antibody
Anticuerpos
Type: Secondary
Antigen: IgM (Mouse) (mouse IgM)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR