Anti-IgA Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9170-1EA 365 EUR
ABCAAB9170-1
Anti-IgA Rabbit Polyclonal Antibody
Anticuerpos
Type: Secondary
Antigen: Mouse IgA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR