Anti-IgG1 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9165-500EA 225 EUR
ABCAAB9165-500
Anti-IgG1 Goat Polyclonal Antibody
Anticuerpos
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR