Anti-IgG2b Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9164-500EA 323 EUR
ABCAAB9164-500
Anti-IgG2b Goat Polyclonal Antibody
Anticuerpos
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR