Anti-IgG2a Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9163-500EA 316 EUR
ABCAAB9163-500
Anti-IgG2a Goat Polyclonal Antibody
Anticuerpos
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR