Anti-IgE Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9162-500EA 340 EUR
ABCAAB9162-500
Anti-IgE Goat Polyclonal Antibody
Anticuerpos
Type: Secondary
Antigen: IgE (Mouse) (mouse IgE)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR