Anti-IgM Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9160-1EA 0 EUR
ABCAAB9160-1
Anti-IgM Goat Polyclonal Antibody
Anticuerpos
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR