Anti-XRCC1 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB9147-50EA 560 EUR
ABCAAB9147-50
Anti-XRCC1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: XRCC1 (X-ray repair complementing defective repair in Chinese hamster cells 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR