Anti-MSH2 Rabbit Polyclonal Antibody

ABCAAB9146-50EA 611 EUR
ABCAAB9146-50
Anti-MSH2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: MSH2 (mutS homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR