Anti-T7 Tag Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB9138-200EA 785 EUR
ABCAAB9138-200
Anti-T7 Tag Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: T7 Tag (T7 tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR