Anti-MMP14 Rabbit Polyclonal Antibody

ABCAAB9122-50EA 611 EUR
ABCAAB9122-50
Anti-MMP14 Rabbit Polyclonal Antibody
Type: Primary
Antigen: MMP14 (matrix metallopeptidase 14 (membrane-inserted))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR