Anti-6X His tag Chicken Polyclonal Antibody

Proveedor: Abcam
ABCAAB9107-250EA 830 EUR
ABCAAB9107-250
Anti-6X His tag Chicken Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: 6X His tag (6X His tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR