Anti-TLR4/MD2 Complex Rat Monoclonal Antibody (Biotin) [clone: MTS510]

Proveedor: Abcam
ABCAAB9105-25EA 831 EUR
ABCAAB9105-25
Anti-TLR4/MD2 Complex Rat Monoclonal Antibody (Biotin) [clone: MTS510]
Anticuerpos
Type: Primary
Antigen: TLR4/MD2 Complex
Clonality: monoclonal
Clone: MTS510
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR