Anti-TLR2 Mouse Monoclonal Antibody [clone: TL2.3]

ABCAAB9102-50EA 645 EUR
ABCAAB9102-50
Anti-TLR2 Mouse Monoclonal Antibody [clone: TL2.3]
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: TL2.3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR