Anti-TLR2 Mouse Monoclonal Antibody [clone: TL2.1]
Proveedor: Abcam
ABCAAB9100-50EA 560 EUR
ABCAAB9100-50
Anti-TLR2 Mouse Monoclonal Antibody [clone: TL2.1]
Anticuerpos
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: TL2.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now